accessibility menu, dialog, popup

UserName

BNFHOOH

CAS RN 1150114-77-4

IUPAC Name: (4-cyano-2-fluorophenyl)boronic acid
Synonyms: 4-cyano-2-fluorophenylboronic acid
Molecular Weight (g/mol): 164.93
Molecular Formula: C7H5BFNO2
InChi Key: HFAUMAIVCYTVQR-UHFFFAOYSA-N
SMILES: OB(O)C1=C(F)C=C(C=C1)C#N
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results