missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-2-fluorobenzeneboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 15437638
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15437638 1 g 1g
15447638 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15437638 Supplier Thermo Scientific Alfa Aesar Supplier No. H61134.03
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

4-Cyano-2-fluorobenzeneboronic acid is used an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyano-2-fluorobenzeneboronic acid is used an active pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1150114-77-4
Molecular Formula C7H5BFNO2
Molecular Weight (g/mol) 164.93
MDL Number MFCD07644472
InChI Key HFAUMAIVCYTVQR-UHFFFAOYSA-N
Synonym 4-cyano-2-fluorophenyl boronic acid, boronic acid, b-4-cyano-2-fluorophenyl, acmc-2099nb, 4-borono-3-fluorobenzonitrile, 4-cyano-2-fluoro-phenylboronic acid, 4-cyano-2-fluorobenzeneboronic acid
PubChem CID 44755209
IUPAC Name (4-cyano-2-fluorophenyl)boronic acid
SMILES OB(O)C1=C(F)C=C(C=C1)C#N

Specifications

Quantity 1 g
Formula Weight 164.93
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Cyano-2-fluorobenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.