accessibility menu, dialog, popup

UserName

CH3(S)H3C(R)OO(S)OHOHOO(R)(R)CH3(S)OH(R)OH(S)OH(S)CH3NH2OO(S)OHOHOO

CAS RN 116355-83-0

IUPAC Name: (2S)-2-(2-{[(5S,6R,7R,9R,11S,16R,18S,19S)-19-amino-6-{[(3S)-3,4-dicarboxybutanoyl]oxy}-11,16,18-trihydroxy-5,9-dimethylicosan-7-yl]oxy}-2-oxoethyl)butanedioic acid
Synonyms: fumonisin B1
Molecular Weight (g/mol): 721.84
Molecular Formula: C34H59NO15
InChi Key: UVBUBMSSQKOIBE-ZWKVXHQASA-N
SMILES: CCCC[C@H](C)[C@@H](OC(=O)C[C@H](CC(O)=O)C(O)=O)[C@@H](C[C@H](C)C[C@@H](O)CCCC[C@@H](O)C[C@H](O)[C@H](C)N)OC(=O)C[C@H](CC(O)=O)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
  • (2)
Quantity 
  • (3)
  • (1)
  • (2)
Industry Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)