missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fumonisins B1 and B2 Mixture 50 μg/mL in Acetonitrile/Water, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Fumonisin B1∣Fumonisin B2 |
| CAS | 116355-83-0, 116355-84-1 |
| Analyte or Component Names | Fumonisin B1∣Fumonisin B2 |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 9001 |
| Solution Type | Mixture |
| Solvent or Matrix | Acetonitrile/Water |
| Industry Type | Environmental Testing |
| Quantity | 5 mL |
| Concentration or Composition (by Analyte or Components) | Fumonisin B1 50 μg/mL∣Fumonisin B2 50 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Ser du en mulighed for forbedring?