CAS RN 1163685-30-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-(1H-Benzo[d]imidazol-2-yl)-3-methyl-4-oxo-1,4-dihydropyridine-2-carboxylic acid (>75%), TRC
CAS: 1163685-30-0 Molecular Formula: C14 H11 N3 O3 Molecular Weight (g/mol): 269.26 Synonym: Rabeprazole Sodium Imp C (EP) IUPAC Name: 1-(1H-benzimidazol-2-yl)-3-methyl-4-oxopyridine-2-carboxylic acid SMILES: CC1=C(N(C=CC1=O)c2nc3ccccc3[nH]2)C(=O)O