missing translation for 'onlineSavingsMsg'
Learn More

1-(1H-Benzo[d]imidazol-2-yl)-3-methyl-4-oxo-1,4-dihydropyridine-2-carboxylic acid (>75%), TRC

Product Code. 30012141
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012141 1 mg 1mg
30004466 5 mg 5mg
30007891 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012141 Supplier Toronto Research Chemicals Supplier No. TRCB2039601MG
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1163685-30-0
Molecular Formula C14 H11 N3 O3
Molecular Weight (g/mol) 269.26
Synonym Rabeprazole Sodium Imp C (EP)
IUPAC Name 1-(1H-benzimidazol-2-yl)-3-methyl-4-oxopyridine-2-carboxylic acid
SMILES CC1=C(N(C=CC1=O)c2nc3ccccc3[nH]2)C(=O)O

Specifications

Analyte or Component Names 1-(1H-Benzimidazol-2-yl)-3-methyl-4-oxo-1,4-dihydropyridine-2-carboxylic acid
Shipping Condition Room Temperature
InChI Formula InChI=1S/C14H11N3O3/c1-8-11(18)6-7-17(12(8)13(19)20)14-15-9-4-2-3-5-10(9)16-14/h2-7H,1H3,(H,15,16)(H,19,20)
Solution Type Neat
Industry Type Forensics
Quantity 1 mg
Formula Weight 269.08
Chemical Name or Material 1-(1H-Benzimidazol-2-yl)-3-methyl-4-oxo-1,4-dihydropyridine-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.