accessibility menu, dialog, popup

UserName

CAS RN 117752-04-2

CH3FO

CAS RN 117752-04-2

IUPAC Name: 2-fluoro-6-methylbenzaldehyde
Synonyms: 2-fluoro-6-methylbenzaldehyde
Molecular Weight (g/mol): 138.14
Molecular Formula: C8H7FO
InChi Key: BGQBJWPZBBMKEI-UHFFFAOYSA-N
SMILES: CC1=C(C=O)C(F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results