missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methylbenzaldehyde, 97%

Product Code. 15431408
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15431408

Brand: Thermo Scientific Alfa Aesar H55228.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 117752-04-2 | C8H7FO | 138.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117752-04-2
Molecular Formula C8H7FO
Molecular Weight (g/mol) 138.14
MDL Number MFCD09258962
InChI Key BGQBJWPZBBMKEI-UHFFFAOYSA-N
PubChem CID 14170517
IUPAC Name 2-fluoro-6-methylbenzaldehyde
SMILES CC1=C(C=O)C(F)=CC=C1

Specifications

Color Colorless to Yellow
Density 1.151 g/mL
Refractive Index 1.525
Quantity 1 g
Synonym 2-fluoro-6-methyl-benzaldehyde, benzaldehyde,2-fluoro-6-methyl, pubchem10067, 3-fluoro-2-formyltoluene, acmc-209xa9, benzaldehyde, 2-fluoro-6-methyl, 2-fluoro-6-methylbenzaldehyde
Formula Weight 138.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Fluoro-6-methylbenzaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.