accessibility menu, dialog, popup

UserName

SBr

CAS RN 118150-25-7

IUPAC Name: 2-[(4-bromophenyl)methyl]thiophene
Synonyms: 2-[(4-bromophenyl)methyl]thiophene
Molecular Weight (g/mol): 253.16
Molecular Formula: C11H9BrS
InChi Key: GGYCIKURJBHUER-UHFFFAOYSA-N
SMILES: BrC1=CC=C(CC2=CC=CS2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results