missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromobenzyl)thiophene, 97%, Thermo Scientific™

Product Code. 10674015
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10674015 5 g Amber Glass Bottle 5g
10113864 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10674015 Supplier Thermo Scientific Maybridge Supplier No. CC70610DE
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 118150-25-7
Molecular Formula C11H9BrS
Molecular Weight (g/mol) 253.157
MDL Number MFCD11841076
InChI Key GGYCIKURJBHUER-UHFFFAOYSA-N
Synonym 2-4-bromobenzyl thiophene, 2-4-bromophenyl methyl thiophene
PubChem CID 15322628
IUPAC Name 2-[(4-bromophenyl)methyl]thiophene
SMILES C1=CSC(=C1)CC2=CC=C(C=C2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 253.16
Percent Purity 97%
Chemical Name or Material 2-(4-Bromobenzyl)thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.