accessibility menu, dialog, popup

UserName

CAS RN 1189-71-5

ClSOONCO

CAS RN 1189-71-5

IUPAC Name: [(chlorosulfonyl)imino]methanone
Synonyms: chlorosulfonyl isocyanate sulfuryl chloride isocyanate
Molecular Weight (g/mol): 141.53
Molecular Formula: CClNO3S
InChi Key: WRJWRGBVPUUDLA-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)N=C=O
Boiling Point 
  • (5)
Physical Form 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)