accessibility menu, dialog, popup

UserName

H2NNONHNOHONNH

CAS RN 119-24-4

IUPAC Name: 4-{[(2-amino-4-oxo-1,4-dihydropteridin-6-yl)methyl]amino}benzoic acid
Synonyms: 4-{[(2-amino-4-oxo-1H-pteridin-6-yl)methyl]amino}benzoic acid
Molecular Weight (g/mol): 312.29
Molecular Formula: C14H12N6O3
InChi Key: JOAQINSXLLMRCV-UHFFFAOYSA-N
SMILES: NC1=NC(=O)C2=NC(CNC3=CC=C(C=C3)C(O)=O)=CN=C2N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results