accessibility menu, dialog, popup

UserName

CAS RN 119-30-2

HOOIHO

CAS RN 119-30-2

IUPAC Name: 2-hydroxy-5-iodobenzoic acid
Synonyms: 2-hydroxy-5-iodobenzoic acid
Molecular Weight (g/mol): 264.02
Molecular Formula: C7H5IO3
InChi Key: SWDNKOFGNPGRPI-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(I)=CC=C1O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)