Learn More
5-Iodosalicylic acid, 97%
Description
2-hydroxy-5-iodobenzoic acid is a reagent in various organic chemical reactions leading to pharmaceutical compounds. It is involved in the synthesis of labeled benzamide analogs for tumor imaging as well as the synthesis of the dye, Congo Red, for biological analyses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-hydroxy-5-iodobenzoic acid is a reagent in various organic chemical reactions leading to pharmaceutical compounds. It is involved in the synthesis of labeled benzamide analogs for tumor imaging as well as the synthesis of the dye, Congo Red, for biological analyses.
Solubility
Solubility in methanol is almost transparent.
Notes
Light sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 194°C to 199°C |
| Quantity | 500 g |
| Beilstein | 2090443 |
| Formula Weight | 264.02 |
| Percent Purity | 97% |
| Chemical Name or Material | 5-Iodosalicylic acid |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.