accessibility menu, dialog, popup

UserName

NH2SOOONOOSOOO(E)

CAS RN 119-72-2

IUPAC Name: 5-amino-2-[(1E)-2-(4-nitro-2-sulfonatophenyl)ethenyl]benzene-1-sulfonate
Synonyms: 5-amino-2-[(1E)-2-(4-nitro-2-sulfonatophenyl)ethenyl]benzenesulfonate
Molecular Weight (g/mol): 398.36
Molecular Formula: C14H10N2O8S2
InChi Key: GHBWBMDGBCKAQU-OWOJBTEDSA-L
SMILES: NC1=CC=C(\C=C\C2=CC=C(C=C2S([O-])(=O)=O)[N+]([O-])=O)C(=C1)S([O-])(=O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results