missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-4'-nitrostilbene-2,2'-disulfonic acid, tech. 80%, Thermo Scientific Chemicals

Product Code. 11436994
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11436994

Brand: Thermo Scientific Alfa Aesar B21467.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

4-Amino-4'-nitrostilbene-2,2'-disulfonic acid serves as an important stilbene intermediate. It acts as a monomer. Further, it is used in the synthesis of a series of direct dyes such as direct lightfast fluorescent yellow 7GL, 6GL, 5GL, 4GL, 3GL and 2GL. In addition to this, it is used to prepare direct yellow DN-FR, orange F3G; yellow 3RLL, green GND and direct Black DN-EX.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-4′-nitrostilbene-2,2′-disulfonic acid serves as an important stilbene intermediate. It acts as a monomer. Further, it is used in the synthesis of a series of direct dyes such as direct lightfast fluorescent yellow 7GL, 6GL, 5GL, 4GL, 3GL and 2GL. In addition to this, it is used to prepare direct yellow DN-FR, orange F3G; yellow 3RLL, green GND and direct Black DN-EX.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-72-2
Molecular Formula C14H12N2O8S2
Molecular Weight (g/mol) 400.376
MDL Number MFCD00035915
InChI Key GHBWBMDGBCKAQU-OWOJBTEDSA-N
Synonym 4-nitro-4'-aminostilbene-2,2'-disulfonic acid, 4-amino-4'-nitro-2,2'-stilbenedisulfonic acid, 4-amino-4'-nitrostilbene-2,2'-disulfonic acid, kyselina 4-amino-4'-nitrostilben-2,2'-disulfonova czech, kyselina 4-nitro-4'-aminostilben-2,2'-disulfonova czech, benzenesulfonic acid, 5-amino-2-2-4-nitro-2-sulfophenyl ethenyl, 2,2'-stilbenedisulfonic acid, 4-amino-4'-nitro, 5-amino-2-2-4-nitro-2-sulphophenyl vinyl benzenesulphonic acid, 5-amino-2-e-2-4-nitro-2-sulfo-phenyl vinyl benzenesulfonic acid, 5-amino-2-e-2-4-nitro-2-sulfophenyl ethenyl benzenesulfonic acid
PubChem CID 5473847
IUPAC Name 5-amino-2-[(E)-2-(4-nitro-2-sulfophenyl)ethenyl]benzenesulfonic acid
SMILES C1=CC(=C(C=C1N)S(=O)(=O)O)C=CC2=C(C=C(C=C2)[N+](=O)[O-])S(=O)(=O)O

Specifications

Melting Point >300°C
Odor Odorless
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 400.38
Percent Purity 80%
Grade Technical
Chemical Name or Material 4-Amino-4'-nitrostilbene-2,2'-disulfonic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.