accessibility menu, dialog, popup

UserName

H3CCH3OO

CAS RN 120-50-3

IUPAC Name: 2-methylpropyl benzoate
Synonyms: isobutyl benzoate
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: KYZHGEFMXZOSJN-UHFFFAOYSA-N
SMILES: CC(C)COC(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)