missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl benzoate, 98%, ACROS Organics™

Product Code. 10565481 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10565481 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10565481 Supplier Thermo Scientific Acros Supplier No. 350670250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 120-50-3
Color Colorless
Density 0.999g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C11H14O2
Linear Formula C6H5C(O)OCHCH(CH3)
Refractive Index 1.4930 to 1.4950 (20°C, 589nm)
MDL Number MFCD00048344
Quantity 25 g
Synonym isobutyl benzoate, benzoic acid, isobutyl ester, benzoic acid, 2-methylpropyl ester, unii-kq6xz9wjii, isobutyl benzoate natural, fema no. 2185, kq6xz9wjii, benzoic acid isobutyl ester, benzoic acid, isobutyl ester 6ci,7ci,8ci, iso-butyl benzoate
InChI Key KYZHGEFMXZOSJN-UHFFFAOYSA-N
SMILES CC(C)COC(=O)C1=CC=CC=C1
IUPAC Name 2-methylpropyl benzoate
Molecular Weight (g/mol) 178.23
PubChem CID 61048
ChEBI CHEBI:87500
Formula Weight 178.23
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material Isobutyl benzoate, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.