accessibility menu, dialog, popup

UserName

RhClRhCl

CAS RN 12092-47-6

IUPAC Name: cyclooctadiene rhodium chloride dimer
Synonyms: cyclooctadiene rhodium chloride dimer
Molecular Weight (g/mol): 493.08
Molecular Formula: C16H24Cl2Rh2
InChi Key: LGARIVPRHUAOKP-MIXQCLKLSA-N
SMILES: [Cl].[Cl].[Rh].[Rh].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)