missing translation for 'onlineSavingsMsg'
Learn More

Chloro(1,5-cyclooctadiene)rhodium(I) dimer, min. 40.8% Rh

Product Code. 10492845 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
5 g
Packaging:
Glass bottle
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10492845 5 g Glass bottle 5g
10266600 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10492845 Supplier Thermo Scientific Acros Supplier No. 211300050
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 12092-47-6 | C16H24Cl2Rh2 | 493.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12092-47-6
Molecular Formula C16H24Cl2Rh2
Molecular Weight (g/mol) 493.08
MDL Number MFCD00012415
InChI Key QSUDXYGZLAJAQU-MIXQCLKLSA-L
Synonym chloro 1,5-cyclooctadiene rhodium i dimer, 1z,5z-cycloocta-1,5-diene; rhodium; dichloride, bis 1,5-cyclooctadiene dirhodium i dichloride, 1,5-cyclooctadiene rhodium chloride dimer, ccris 5036, di-?-chlorobis 1,2,5,6-?-1,5-cyclooctadiene diiridium, rhodium-1,5-cyclooctadiene chloride, 1,5-cyclooctadiene-iridium i chloride dimer, chloro 1,5-cyclooctadiene iridium i dimer
PubChem CID 6436379
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;rhodium;dichloride
SMILES [Cl-].[Cl-].[Rh+].[Rh+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1

Specifications

Melting Point 256.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: 0.34g/L
Formula Weight 493.08
Percent Purity ≥40.8% (RH)
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Chloro(1, 5-cyclooctadiene)rhodium(I) dimer, min. 40.8% Rh

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.