CAS RN 1212021-62-9
CAS RN 1212021-62-9
IUPAC Name:
[3-cyano-5-(trifluoromethyl)phenyl]boronic acid
Synonyms:
3-cyano-5-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol):
214.94
Molecular Formula:
C8H5BF3NO2
InChi Key:
CTJAWCWPRYNDDQ-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC(=CC(=C1)C(F)(F)F)C#N
1
–
1
of
1
results
1
3-Cyano-5-(trifluoromethyl)benzeneboronic acid, 97%, Thermo Scientific™
CAS: 1212021-62-9 Molecular Formula: C8H5BF3NO2 Molecular Weight (g/mol): 214.94 MDL Number: MFCD03095102 InChI Key: CTJAWCWPRYNDDQ-UHFFFAOYSA-N Synonym: 3-cyano-5-trifluoromethyl phenyl boronic acid,3-cyano-5-trifluoromethyl phenylboronic acid,acmc-209a9l,3-cyano-5-trifluoromethyl phenyl boronicacid PubChem CID: 56776695 IUPAC Name: [3-cyano-5-(trifluoromethyl)phenyl]boronic acid SMILES: OB(O)C1=CC(=CC(=C1)C(F)(F)F)C#N