accessibility menu, dialog, popup

UserName

NH(S)OHNOHOCH3

CAS RN 1218-34-4

IUPAC Name: 2-acetamido-3-(1H-indol-3-yl)propanoic acid
Synonyms: N-acetyltryptophan
Molecular Weight (g/mol): 246.27
Molecular Formula: C13H14N2O3
InChi Key: DZTHIGRZJZPRDV-UHFFFAOYNA-N
SMILES: CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (6)
Quantity 
  • (1)
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results