CAS RN 122-42-9
IUPAC Name:
propan-2-yl N-phenylcarbamate
Synonyms:
isopropyl phenyl carbamate
Molecular Weight (g/mol):
179.22
Molecular Formula:
C10H13NO2
InChi Key:
VXPLXMJHHKHSOA-UHFFFAOYSA-N
SMILES:
CC(C)OC(=O)NC1=CC=CC=C1
1
–
3
of
3
results
2
Isopropyl N-phenylcarbamate, 96%, Thermo Scientific™
CAS: 122-42-9 Molecular Formula: C10H13NO2 Molecular Weight (g/mol): 179.219 MDL Number: MFCD00026382 InChI Key: VXPLXMJHHKHSOA-UHFFFAOYSA-N Synonym: propham,isopropyl n-phenylcarbamate,isopropyl phenylcarbamate,triherbide,agermin,collavin,prophame,tuberit,tuberite,birgin PubChem CID: 24685 ChEBI: CHEBI:82035 IUPAC Name: propan-2-yl N-phenylcarbamate SMILES: CC(C)OC(=O)NC1=CC=CC=C1