accessibility menu, dialog, popup

UserName

CH3NONNHF

CAS RN 1223001-53-3

IUPAC Name: 1-(4-fluoro-3-methylphenyl)-1H,2H,7H-pyrrolo[2,3-h]1,6-naphthyridin-2-one
Synonyms: 1-(4-fluoro-3-methylphenyl)-7H-pyrrolo[2,3-h]1,6-naphthyridin-2-one
Molecular Weight (g/mol): 293.3
Molecular Formula: C17H12FN3O
InChi Key: WQNRDXHKVSKUPI-UHFFFAOYSA-N
SMILES: CC1=C(F)C=CC(=C1)N1C(=O)C=CC2=CN=C3NC=CC3=C12
Grade 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results