missing translation for 'onlineSavingsMsg'
Learn More

STK16-IN-1, MedChemExpress

Product Code. 16499846
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499846 50 mg 50mg
16446037 5 mg 5mg
16432716 25 mg 25mg
16423318 10 mg 10mg
16414566 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16499846 Supplier Medchem Express Supplier No. HY101270

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress STK16-IN-1 is a STK16 kinase inhibitor with an IC50 of 295 nM.

STK16-IN-1 is a STK16 kinase inhibitor with an IC50 of 295 nM.

Specifications

Chemical Name or Material STK16-IN-1
CAS 1223001-53-3
Color White
Purity Grade Notes Research
Molecular Formula C17H12FN3O
Quantity 50 mg
Solubility Information DMSO : 15 mg/mL (51.14 mM; Need ultrasonic and warming)
SMILES FC1=CC=C(N2C(C(C=CC2=O)=CN3)=C4C3=NC=C4)C=C1C
Molecular Weight (g/mol) 293.3
Formula Weight 293.3
Percent Purity 99.49%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.