accessibility menu, dialog, popup

UserName

CH3OCH3CH3HO

CAS RN 123-42-2

IUPAC Name: 4-hydroxy-4-methylpentan-2-one
Synonyms: diacetone alcohol diacetonyl alcohol diacetonalkohol dimethyl acetonyl carbinol diacetone-alcool
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: SWXVUIWOUIDPGS-UHFFFAOYSA-N
SMILES: CC(=O)CC(C)(C)O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (3)
  • (4)
  • (4)
  • (1)