missing translation for 'onlineSavingsMsg'
Learn More

Diacetone Alcohol, TRC

Product Code. 30179674
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179674 25 mL 25mL
30178993 500 mL 500mL
30179118 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179674 Supplier Toronto Research Chemicals Supplier No. TRCD67990025ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-42-2
Molecular Formula C6 H12 O2
Molecular Weight (g/mol) 116.16
Synonym 2-Pentanone, 4-hydroxy-4-methyl- (8CI, 9CI, ACI), 4-Hydroxy-4-methyl-2-pentanone (ACI), 2-Hydroxy-2-methyl-4-pentanone, 2-Methyl-2-pentanol-4-one, 2-Methyl-4-oxo-2-pentanol, 2-Methyl-4-oxopentan-2-ol, 4-Hydroxy-2-keto-4-methylpentane, 4-Hydroxy-4-methyl-2-oxopentane, 4-Methyl-4-hydroxy-2-pentanone, Acetonyldimethylcarbinol, Diacetone alcohol, Diketone alcohol, NSC 9005, Pyranton A, Tyranton
IUPAC Name 4-hydroxy-4-methylpentan-2-one
SMILES CC(=O)CC(C)(C)O

Specifications

Quantity 25 mL
InChI Formula InChI=1S/C6H12O2/c1-5(7)4-6(2,3)8/h8H,4H2,1-3H3
Formula Weight 116.0837
Chemical Name or Material 4-Hydroxy-4-methyl-2-pentanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.