accessibility menu, dialog, popup

UserName

CAS RN 123-78-4

H3C(R)OH(S)H2NOH(E)

CAS RN 123-78-4

IUPAC Name: (2S,3R,4E)-2-aminooctadec-4-ene-1,3-diol
Synonyms: sphingosine (-)-D-erythro-sphingosine 2-amino-4-octadecene-1,3-diol sphingoid Sph 4-trans-sphingenine sphingenine 4-sphingenine (2S,3R)-sphingosine sphingosine (substance)
Molecular Weight (g/mol): 299.5
Molecular Formula: C18H37NO2
InChi Key: WWUZIQQURGPMPG-KRWOKUGFSA-N
SMILES: CCCCCCCCCCCCC\C=C\[C@@H](O)[C@@H](N)CO
Molecular Weight (g/mol) 
  • (4)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (4)
  • (2)
Physical Form 
  • (1)
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Color 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (4)