accessibility menu, dialog, popup

UserName

H3CCH3OCH3O

CAS RN 123-92-2

IUPAC Name: 3-methylbutyl acetate
Synonyms: banana oil fema number 2055 common amyl acetate amyl acetate, common acetate de 3-methylbutyle pear oil amylacetic ester β-methyl butyl acetate acetate d'isoamyle isoamyl acetate (substance)
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: MLFHJEHSLIIPHL-UHFFFAOYSA-N
SMILES: CC(C)CCOC(C)=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
Boiling Point 
  • (9)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (9)
Percent Purity 
  • (3)
  • (3)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (9)
  • (3)
  • (3)
  • (9)
  • (3)
  • (3)
  • (2)
  • (3)
  • (7)
  • (3)
  • (3)
  • (1)