missing translation for 'onlineSavingsMsg'
Learn More

Iso-Amyl Acetate, Extra Pure, SLR, mix of 2- & 3-methylbutyl acetate, Fisher Chemical™

Product Code. 10776471 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10776471 500 mL 500mL
10375500 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10776471 Supplier Fisher Chemical Supplier No. A/6880/08
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 123-92-2
Molecular Formula C7H14O2
MDL Number 8946

Specifications

Melting Point -78°C
Color Colorless
Boiling Point 142°C
Packaging Amber glass bottle
Quantity 500 mL
Vapor Density 4.49
Formula Weight 130.19g/mol
Vapor Pressure 4mmHg at 20°C
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.