accessibility menu, dialog, popup

UserName

CH3CH3CH3CCH3CH3CH3CH3CH3CCH3CH3ClClClClIrIr

CAS RN 12354-84-6

IUPAC Name: diiridium(3+) bis(pentamethylcyclopenta-2,4-dien-1-ide) tetrachloride
Synonyms: diiridium(3+) bis(pentamethylcyclopenta-2,4-dien-1-ide) tetrachloride
Molecular Weight (g/mol): 796.69
Molecular Formula: C20H30Cl4Ir2
InChi Key: MMAGMBCAIFVRGJ-UHFFFAOYSA-J
SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Ir+3].[Ir+3].C[C-]1C(C)=C(C)C(C)=C1C.C[C-]1C(C)=C(C)C(C)=C1C
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)