missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer

Product Code. 15457005
Change view
Click to view available options
Quantity:
500 mg
2 g
Unit Size:
2g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15457005 500 mg 500mg
15447005 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15457005 Supplier Thermo Scientific Alfa Aesar Supplier No. 045565.ME
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 12354-84-6 | C20H30Cl4Ir2 | 796.694 g/mol

Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer is used as a precursor to catalysts for the asymmetric transfer hydrogenation of ketones. It is a catalyst for greener amine synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12354-84-6
Molecular Formula C20H30Cl4Ir2
Molecular Weight (g/mol) 796.694
MDL Number MFCD00075435
InChI Key MMAGMBCAIFVRGJ-UHFFFAOYSA-J
Synonym unii-6oy4uuc534, dichloro pentamethylcyclopentadienyl iridium iii dimer, pentamethylcyclopentadienyl iridium iii dichloride dimer, pentamethylcyclopentadienyliridium dichloride, pentamethylcyclopentadienyl iridium dichloride dimer, pentamethylcyclopentadienyliridium iii dichloride dimer, eta5-pentamethylcyclopentadienyl iridium dichloride dimer, pentamethylcyclopentadienyliridium iii dichloride dimer mi, bis mu-chloro chloro eta5-pentamethylcyclopentadienyl iridium, iridium, di-mu-chlorodichlorobis 1,2,3,4,5-eta-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl di
PubChem CID 76030743
IUPAC Name iridium(3+);1,2,3,4,5-pentamethylcyclopenta-1,3-diene;tetrachloride
SMILES C[C-]1C(=C(C(=C1C)C)C)C.C[C-]1C(=C(C(=C1C)C)C)C.[Cl-].[Cl-].[Cl-].[Cl-].[Ir+3].[Ir+3]

Specifications

Quantity 500 mg
Formula Weight 796.67
Chemical Name or Material Dichloro (pentamethylcyclopentadienyl)iridium (III) dimer

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.