accessibility menu, dialog, popup

UserName

CAS RN 124-17-4

IUPAC Name: 2-(2-butoxyethoxy)ethyl acetate
Synonyms: buteth-2 acetate diethylene glycol monobutyl ether acetate
Molecular Weight (g/mol): 204.27
Molecular Formula: C10H20O4
InChi Key: VXQBJTKSVGFQOL-UHFFFAOYSA-N
SMILES: CCCCOCCOCCOC(C)=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)