CAS RN 124-94-7
CAS RN 124-94-7
IUPAC Name:
(1S,2R,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,2,10-trihydroxy-1-(2-hydroxyacetyl)-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Synonyms:
triamcinolone nasacort kenalog nasacort aq aristocort fougera azmacort vetalog triderm kenacort
Molecular Weight (g/mol):
394.44
Molecular Formula:
C21H27FO6
InChi Key:
GFNANZIMVAIWHM-OBYCQNJPSA-N
SMILES:
C[C@]12C[C@H](O)[C@@]3(F)[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1C[C@@H](O)[C@]2(O)C(=O)CO
1
–
4
of
4
results
1
Thermo Scientific Chemicals Triamcinolone
CAS: 124-94-7 Molecular Formula: C21H27FO6 Molecular Weight (g/mol): 394.43 InChI Key: GFNANZIMVAIWHM-OBYCQNJPSA-N PubChem CID: 31307 ChEBI: CHEBI:9667
2
Triamcinolone, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
3
Triamcinolone, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4
Triamcinolone, MedChemExpress
MedChemExpress Triamcinolone is a long-acting synthetic corticosteroid.