accessibility menu, dialog, popup

UserName

CH3OOHOFF

CAS RN 124728-45-6

IUPAC Name: 4-ethoxy-2,3-difluorobenzoic acid
Synonyms: 4-ethoxy-2,3-difluorobenzoic acid
Molecular Weight (g/mol): 202.16
Molecular Formula: C9H8F2O3
InChi Key: KMFHBAZUQFJVKK-UHFFFAOYSA-N
SMILES: CCOC1=C(F)C(F)=C(C=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results