missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-2,3-difluorobenzoic acid, 98%, Thermo Scientific™

Product Code. 15483367
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15483367 250 mg 250mg
11342049 1 g 1g
15473367 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15483367 Supplier Thermo Scientific Alfa Aesar Supplier No. H26089.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124728-45-6
Molecular Formula C9H8F2O3
Molecular Weight (g/mol) 202.157
MDL Number MFCD02258849
InChI Key KMFHBAZUQFJVKK-UHFFFAOYSA-N
PubChem CID 2769400
IUPAC Name 4-ethoxy-2,3-difluorobenzoic acid
SMILES CCOC1=C(C(=C(C=C1)C(=O)O)F)F

Specifications

Melting Point 200°C to 203°C
Quantity 250 mg
Synonym 2,3-difluoro-4-ethoxybenzoic acid, benzoic acid, 4-ethoxy-2,3-difluoro, acmc-20aari, 4-carboxy-2,3-difluorophenetole, benzoic acid,4-ethoxy-2,3-difluoro, benzoic acid, 4-ethoxy-2,3-difluoro-9ci
Formula Weight 202.15
Percent Purity 98%
Chemical Name or Material 4-Ethoxy-2,3-difluorobenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.