accessibility menu, dialog, popup

UserName

CAS RN 125-13-3

OHOHNHO

CAS RN 125-13-3

IUPAC Name: 3,3-bis(4-hydroxyphenyl)-2,3-dihydro-1H-indol-2-one
Synonyms: oxyphenisatin oxyphenisatinum oxifenisatina [inn-spanish] phenolisatin diphenolisatin dihydroxyphenoloxindol 4,4'-dihydroxydiphenylisatin veripaque isolax lavema
Molecular Weight (g/mol): 317.34
Molecular Formula: C20H15NO3
InChi Key: SJDACOMXKWHBOW-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C1(C(=O)NC2=CC=CC=C12)C1=CC=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results