accessibility menu, dialog, popup

UserName

CAS RN 125-84-8

IUPAC Name: 3-(4-aminophenyl)-3-ethylpiperidine-2,6-dione
Synonyms: aminoglutethimide aminoglutethimide preparation aminoglutethimidum AGT orimeten ciba vision brand of aminoglutethimide aminoglutetimida 氨鲁米特 aminoglutethimide (substance) elipten
Molecular Weight (g/mol): 232.28
Molecular Formula: C13H16N2O2
InChi Key: ROBVIMPUHSLWNV-UHFFFAOYNA-N
SMILES: CCC1(CCC(=O)NC1=O)C1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results