accessibility menu, dialog, popup

UserName

CAS RN 126519-89-9

IUPAC Name: 4-[2-(trimethoxysilyl)ethyl]benzene-1-sulfonyl chloride
Synonyms: 4-[2-(trimethoxysilyl)ethyl]benzenesulfonyl chloride
Molecular Weight (g/mol): 324.85
Molecular Formula: C11H17ClO5SSi
InChi Key: NYIDSUMRGUILGR-UHFFFAOYSA-N
SMILES: CO[Si](CCC1=CC=C(C=C1)S(Cl)(=O)=O)(OC)OC
Boiling Point 
  • (3)
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)