missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorosulfonylphenyl)ethyltrimethoxysilane, 50% solution in dichloromethane

Product Code. 10603172 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10603172 25 g Glass bottle 25g
10555491 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10603172 Supplier Thermo Scientific Acros Supplier No. 337470250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 126519-89-9 | C11H17ClO5SSi | 324.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126519-89-9 , 75-09-2
Molecular Formula C11H17ClO5SSi
Molecular Weight (g/mol) 324.85
MDL Number MFCD00054774
InChI Key NYIDSUMRGUILGR-UHFFFAOYSA-N
Synonym 2-4-chlorosulfonylphenyl ethyltrimethoxysilane, 4-2-trimethoxysilylethyl benzenesulfonyl chloride, 4-2-trimethoxysilyl ethyl benzene-1-sulfonyl chloride, benzenesulfonylchloride, 4-2-trimethoxysilyl ethyl, 2-4-chlorosulfonylphenyl ethyltrimethoxysilane in methylene chloride, has free sulfonic acid, 4-2-trimethoxysilyl ethyl benzenesulfonyl chloride, acmc-20ete8, 4-chlorosulfonyl phenethyltrimethoxysilane
PubChem CID 2733807
IUPAC Name 4-(2-trimethoxysilylethyl)benzenesulfonyl chloride
SMILES CO[Si](CCC1=CC=C(C=C1)S(Cl)(=O)=O)(OC)OC

Specifications

CAS Min % 45.0
CAS Max % 55.0
Color Yellow
Boiling Point >40.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: reacts
Formula Weight 324.85
Percent Purity 45 to 55% (solids)
Physical Form Solution
Chemical Name or Material 2-(4-Chlorosulfonylphenyl)ethyltrimethoxysilane
Show More Show Less
compliance-icons compliance-icons
Produktidentifikator
  • 2-(4-Chlorosulfonylphenyl)ethyltrimethoxysilane, 50% solution in dichloromethane
Signalord
  • Danger
Farekategori
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Faresætning
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H336-May cause drowsiness or dizziness.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.