accessibility menu, dialog, popup

UserName

NNO

CAS RN 127404-22-2

IUPAC Name: 3-(1H-imidazol-1-yl)benzaldehyde
Synonyms: 3-(imidazol-1-yl)benzaldehyde
Molecular Weight (g/mol): 172.19
Molecular Formula: C10H8N2O
InChi Key: CGTQCZGDVDTRDI-UHFFFAOYSA-N
SMILES: O=CC1=CC(=CC=C1)N1C=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results