accessibility menu, dialog, popup

UserName

CAS RN 129931-45-9

IUPAC Name: 3-chloro-2-fluoro-5-(trifluoromethyl)benzoate
Synonyms: 3-chloro-2-fluoro-5-(trifluoromethyl)benzoate
Molecular Weight (g/mol): 241.55
Molecular Formula: C8H2ClF4O2
InChi Key: UINXNXRHKMRASA-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(=CC(Cl)=C1F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results