missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-fluoro-5-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 11456686
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456686 5 g 5g
11440318 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11456686 Supplier Thermo Scientific Alfa Aesar Supplier No. B23839.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Chloro-2-fluoro-5-(trifluoromethyl)benzoic acid is used as a pharmaceutical intermediate.

Solubility
Soluble in most organic solvents, basic aqueous solutions.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129931-45-9
Molecular Formula C8H3ClF4O2
Molecular Weight (g/mol) 242.554
MDL Number MFCD01631509
InChI Key UINXNXRHKMRASA-UHFFFAOYSA-N
Synonym 3-chloro-2-fluoro-5-trifluoromethyl benzoic acid, rarechem al be 0499, 3-chloro-2-fluoro-5-trifluoromethyl-benzoic acid, pubchem4559, acmc-20a41y, 2-fluoro-3-chloro-5-trifluoromethyl-benzoic acid, benzoic acid,3-chloro-2-fluoro-5-trifluoromethyl, 3-chloro-2-fluoro-5-trifluoromethyl benzoicacid
PubChem CID 2773751
IUPAC Name 3-chloro-2-fluoro-5-(trifluoromethyl)benzoic acid
SMILES C1=C(C=C(C(=C1Cl)F)C(=O)O)C(F)(F)F

Specifications

Melting Point 89°C to 91°C
Quantity 5 g
Solubility Information Soluble in most organic solvents,basic aqueous solutions.
Formula Weight 242.56
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-fluoro-5-(trifluoromethyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.