CAS RN 130676-64-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(S)-3-Hexyl-5,6-dihydro-6-undecyl-2H-pyran-2-one, TRC
CAS: 130676-64-1 Molecular Formula: C22 H40 O2 Molecular Weight (g/mol): 336.55 Synonym: (S)-3-Hexyl-5,6-dihydro-6-undecyl-2H-pyran-2-one (9CI) IUPAC Name: (2S)-5-hexyl-2-undecyl-2,3-dihydropyran-6-one SMILES: CCCCCCCCCCC[C@H]1CC=C(CCCCCC)C(=O)O1
2
(S)-3-Hexyl-5,6-dihydro-6-undecyl-2H-pyran-2-one, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.