CAS RN 132292-17-2
CAS RN 132292-17-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6-(4-Methoxyphenyl)-2-naphthoic Acid, TRC
CAS: 132292-17-2 Molecular Formula: C18H14O3 Molecular Weight (g/mol): 278.3 Synonym: 2-(4-Methoxyphenyl)naphthalene-6-carboxylic Acid,6-(4-Methoxyphenyl)-2-naphthalenecarboxylic Acid IUPAC Name: 6-(4-methoxyphenyl)naphthalene-2-carboxylic acid SMILES: COc1ccc(cc1)c2ccc3cc(ccc3c2)C(=O)O