accessibility menu, dialog, popup

UserName

CH3OOHFFFF

CAS RN 1323955-61-8

IUPAC Name: 2-fluoro-5-methyl-4-(trifluoromethyl)benzoic acid
Synonyms: 2-fluoro-5-methyl-4-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 222.14
Molecular Formula: C9H6F4O2
InChi Key: FRUQXARTJIZTHE-UHFFFAOYSA-N
SMILES: CC1=C(C=C(F)C(=C1)C(O)=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results