accessibility menu, dialog, popup

UserName

OHO(S)NHOO

CAS RN 132684-59-4

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-phenylbutanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-phenylbutanoic acid
Molecular Weight (g/mol): 401.46
Molecular Formula: C25H23NO4
InChi Key: CIHPCIUGLIZADU-QHCPKHFHSA-N
SMILES: OC(=O)[C@H](CCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results