missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-homophenylalanine, 95%, Thermo Scientific Chemicals

Product Code. 15418447
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15418447 5 g 5g
15428447 250 mg 250mg
15408447 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15418447 Supplier Thermo Scientific Alfa Aesar Supplier No. H51973.06
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 132684-59-4 | Molecular Formula: C25H23NO4 | Molecular Weight: 401.46 g/mol

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132684-59-4
Molecular Formula C25H23NO4
Molecular Weight (g/mol) 401.46
MDL Number MFCD00077046
InChI Key CIHPCIUGLIZADU-QHCPKHFHSA-N
Synonym fmoc-l-homophenylalanine, fmoc-homophe-oh, fmoc-hophe-oh, fmoc-l-homophenyl-ala, fmoc-l-homophe, fmoc-l-hphe-oh, n-fmoc-l-homophenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-phenylbutanoic acid, s-2-fmoc-amino-4-phenylbutyric acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-phenylbutanoic acid
PubChem CID 2761467
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-phenylbutanoic acid
SMILES OC(=O)[C@H](CCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Quantity 5 g
Beilstein 4847669
Solubility Information Slightly soluble in water.
Formula Weight 401.46
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-homophenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.