accessibility menu, dialog, popup

UserName

CAS RN 133053-19-7

H3CNCH3NOH3CONHOHN

CAS RN 133053-19-7

IUPAC Name: 3-{1-[3-(dimethylamino)propyl]-5-methoxy-1H-indol-3-yl}-4-(1H-indol-3-yl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 3-{1-[3-(dimethylamino)propyl]-5-methoxyindol-3-yl}-4-(1H-indol-3-yl)-1H-pyrrole-2,5-dione
Molecular Weight (g/mol): 442.52
Molecular Formula: C26H26N4O3
InChi Key: LLJJDLHGZUOMQP-UHFFFAOYSA-N
SMILES: COC1=CC=C2N(CCCN(C)C)C=C(C2=C1)C1=C(C(=O)NC1=O)C1=CNC2=CC=CC=C12
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (5)
  • (1)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)