missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Go 6983

Product Code. 18074512
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18074512 1 mg 1mg
18044222 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18074512 Supplier R&D Systems™ Supplier No. 2285/1
  • ONLINE EXCLUSIVE PRICE
    195.41 €
    174.30 € / 1mg
    Save 21.11 €
    10% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Broad spectrum PKC inhibitor

Broad spectrum protein kinase C (PKC) inhibitor (IC50 values are 7, 7, 6, 10, 60 and 20000 nM for PKCα, PKCβ, PKCγ, PKCδ, PKCζ and PKCμ respectively). Displays cardioprotective properties; reduces polymorphonuclear leukocyte adherence and infiltration following myocardial ischemia/reperfusion injury. Optimizes naïve human pluripotent stem cell growth and viability following naïve cell derivation from primed ESCs and iPSCs using naïve human stem cell medium (NHSM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133053-19-7
Molecular Formula C26H26N4O3

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 442.51
Purity >98%
Molecular Weight (g/mol) 442.519
Chemical Name or Material Go 6983
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.